Få mere at vide
Fexofenadinone, TRC
Artikelnummer. 30006929
Skift visning
Mængde:
25 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006929 25 mg
30004182 1 g
30004109 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006929 Leverandør Toronto Research Chemicals Leverandørnr. TRCF32252025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76811-98-8
Molekylær formel C32 H37 N O4
Molekylvægt (g/mol) 499.64
Synonym 2-[4-[4-[4-(Hydroxydiphenylmethyl)piperidin-1-yl]butanoyl]phenyl]-2-methylpropanoic Acid, Fexofenadine Hydrochloride Imp. A (EP), Ketofexofenadine
IUPAC navn 2-[4-[4-[4-[hydroxy(diphenyl)methyl]piperidin-1-yl]butanoyl]phenyl]-2-methylpropanoic acid
SMIL CC(C)(C(=O)O)c1ccc(cc1)C(=O)CCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4
Tekniske data
Kemisk navn eller materiale Ketofexofenadine
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C32H37NO4/c1-31(2,30(35)36)25-17-15-24(16-18-25)29(34)14-9-21-33-22-19-28(20-23-33)32(37,26-10-5-3-6-11-26)27-12-7-4-8-13-27/h3-8,10-13,15-18,28,37H,9,14,19-23H2,1-2H3,(H,35,36)
Formel vægt 499.2723
Procent renhed >95%