missing translation for 'onlineSavingsMsg'
Learn More

Fenticonazole nitrate, Thermo Scientific™

637.00 DKK - 1955.00 DKK

Chemical Identifiers

CAS 73151-29-8
Molecular Formula C24H21Cl2N3O4S
Molecular Weight (g/mol) 518.41
MDL Number 00941391
InChI Key FJNRUWDGCVDXLU-UHFFFAOYNA-N
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-{[4-(phenylsulfanyl)phenyl]methoxy}ethyl]-1H-imidazole; nitric acid
SMILES O[N+]([O-])=O.ClC1=CC=C(C(CN2C=CN=C2)OCC2=CC=C(SC3=CC=CC=C3)C=C2)C(Cl)=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17505555
View Documents
Thermo Scientific Acros
468792500
250 mg
637.00 DKK
250mg
Please to purchase this item. Need a web account? Register with us today!
17595545
View Documents
Thermo Scientific Acros
468790010
1 g
1955.00 DKK
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

Health hazard

Exclamation mark

missing translation for 'einecsNumber' : 277-302-6

missing translation for 'rtecsNumber' : NI4780000

Recommended Storage : Normal conditions

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fenticonazole nitrate, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.