Få mere at vide
Fenpiverinium Bromide, TRC
Artikelnummer. 30008455
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008455 10 mg
30009308 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30008455 Leverandør Toronto Research Chemicals Leverandørnr. TRCF24900010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 125-60-0
Molekylær formel C22 H29 N2 O . Br
Molekylvægt (g/mol) 417.38
Synonym Piperidinium, 1-(4-amino-4-oxo-3,3-diphenylbutyl)-1-methyl-, bromide (1:1), 1-(3-Carbamoyl-3,3-diphenylpropyl)-1-methylpiperidinium bromide (6CI,7CI), Piperidinium, 1-(3-carbamoyl-3,3-diphenylpropyl)-1-methyl-, bromide (8CI), Piperidinium, 1-(4-amino-4-oxo-3,3-diphenylbutyl)-1-methyl-, bromide (9CI), 12494Hoechst, 2,2-Diphenyl-2-(2-piperidinoethyl)acetamide methyl bromide, 2-(Diphenylpiperidinoethyl)acetamide methobromide, 2-(Diphenylpiperidinoethyl)acetamide methyl bromide, Baralgin-Amid, Baralgin-Amide, Bulgar amide, Fenpipramide methobromide, Fenpiverinium bromide, Hoechst 12494, Resantin, U 0385, α,α-Diphenyl-1-piperidinebutyramide methobromide, α,α-Diphenyl-γ-piperidylbutyramide methobromide
IUPAC navn 4-(1-methylpiperidin-1-ium-1-yl)-2,2-diphenylbutanamide;bromide
SMIL [Br-].C[N+]1(CCC(C(=O)N)(c2ccccc2)c3ccccc3)CCCCC1
Tekniske data
Kemisk navn eller materiale Fenpiverinium Bromide
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C22H28N2O.BrH/c1-24(16-9-4-10-17-24)18-15-22(21(23)25,19-11-5-2-6-12-19)20-13-7-3-8-14-20;/h2-3,5-8,11-14H,4,9-10,15-18H2,1H3,(H-,23,25);1H
Formel vægt 416.1463
Procent renhed >95%