Få mere at vide
Fenbuconazole-lactone B R-9130, TRC
Artikelnummer. 30456243
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30456243 10 mg
30452948 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30456243 Leverandør Toronto Research Chemicals Leverandørnr. TRCT76773010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 146887-38-9
Molekylær formel C19 H16 Cl N3 O2
Molekylvægt (g/mol) 353.8
Synonym 2(3H)-Furanone, 5-(4-chlorophenyl)dihydro-3-phenyl-3-(1H-1,2,4-triazol-1-ylmethyl)-, trans-, rel-(3R,5R)-5-(4-Chlorophenyl)dihydro-3-phenyl-3-(1H-1,2,4-triazol-1-ylmethyl)-2(3H)-furanone, RH 9129
IUPAC navn (3S,5S)-5-(4-chlorophenyl)-3-phenyl-3-(1,2,4-triazol-1-ylmethyl)oxolan-2-one
SMIL Clc1ccc(cc1)[C@@H]2C[C@](Cn3cncn3)(C(=O)O2)c4ccccc4
Tekniske data
Kemisk navn eller materiale Fenbuconazole-lactone B RH-9130
Mængde 10 mg
InChI formel InChI=1S/C19H16ClN3O2/c20-16-8-6-14(7-9-16)17-10-19(18(24)25-17,11-23-13-21-12-22-23)15-4-2-1-3-5-15/h1-9,12-13,17H,10-11H2/t17-,19+/m0/s1
Formel vægt 353.0931