missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Febantel, Thermo Scientific™
652.00 DKK - 2310.00 DKK
Chemical Identifiers
CAS | 58306-30-2 |
---|---|
Molecular Formula | C20H22N4O6S |
Molecular Weight (g/mol) | 446.48 |
MDL Number | 01738527 |
InChI Key | HMCCXLBXIJMERM-UHFFFAOYSA-N |
IUPAC Name | methyl N-{[(methoxycarbonyl)imino]({[2-(2-methoxyacetamido)-4-(phenylsulfanyl)phenyl]amino})methyl}carbamate |
SMILES | COCC(=O)NC1=CC(SC2=CC=CC=C2)=CC=C1NC(NC(=O)OC)=NC(=O)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17312501
|
Thermo Scientific Acros
468780010 |
1 g |
652.00 DKK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17322501
|
Thermo Scientific Acros
468780050 |
5 g |
2310.00 DKK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
58306-30-2 | |
446.48 | |
HMCCXLBXIJMERM-UHFFFAOYSA-N | |
COCC(=O)NC1=CC(SC2=CC=CC=C2)=CC=C1NC(NC(=O)OC)=NC(=O)OC |
C20H22N4O6S | |
01738527 | |
methyl N-{[(methoxycarbonyl)imino]({[2-(2-methoxyacetamido)-4-(phenylsulfanyl)phenyl]amino})methyl}carbamate |
Specifications
Glass Bottle | |
Available | |
C20H22N4O6S | |
White to Light Yellow | |
3973 | |
HMCCXLBXIJMERM-UHFFFAOYSA-N | |
methyl N-{[(methoxycarbonyl)imino]({[2-(2-methoxyacetamido)-4-(phenylsulfanyl)phenyl]amino})methyl}carbamate | |
446.48 |
58306-30-2 | |
Powder/Crystals | |
01738527 | |
129°C to 130°C | |
Insoluble in water | |
COCC(=O)NC1=CC(SC2=CC=CC=C2)=CC=C1NC(NC(=O)OC)=NC(=O)OC | |
446.48 | |
Febantel |
Safety and Handling
GHS07: Exclamation mark
missing translation for 'shelfLife' : 5 years
missing translation for 'einecsNumber' : 261-205-0
missing translation for 'rtecsNumber' : FB3955000
Recommended Storage : Freezer -20°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Febantel, Thermo Scientific™