Få mere at vide
Tocris Bioscience™ Fatostatin A
Artikelnummer. 18054542
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18054542 50 mg
18015332 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18054542 Leverandør Tocris Bioscience™ Leverandørnr. 4444/50
Cell permeable inhibitor of SREBP activation
Inhibitor of sterol regulatory element binding protein (SREBP); impairs the activation of SREBP-1 and SREBP-2. Exhibits antiproliferative effects in DU 145 cells independently of IGF-1 signaling (IC50 = 0.1 μM); reverses hyperglycemia in diabetic (ob/ob) mice. Cell permeable.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 298197-04-3
Molekylær formel C18H19BrN2S
Tekniske data
Kemisk navn eller materiale Fatostatin A
Mængde 50 mg
Formel vægt 375.33
Renhed >97%
Molekylvægt (g/mol) 375.328