Få mere at vide
exo-2,3-Norbornanedicarboxylic Acid Monoamide, TRC
Artikelnummer. 30419468
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30419468 100 mg
30418505 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30419468 Leverandør Toronto Research Chemicals Leverandørnr. TRCN661255100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1365987-23-0
Molekylær formel C9H13NO3
Molekylvægt (g/mol) 183.2
Synonym (1R,2S,3R,4S)-rel-3-(Aminocarbonyl)bicyclo[2.2.1]heptane-2-carboxylic Acid
IUPAC navn (1S,2R,3S,4R)-3-carbamoylbicyclo[2.2.1]heptane-2-carboxylic acid
SMIL O=C([C@]1([H])[C@@]2([H])CC[C@]([H])(C2)[C@]1([H])C(N)=O)O
Tekniske data
Kemisk navn eller materiale exo-2,3-Norbornanedicarboxylic Acid Monoamide
Mængde 100 mg
InChI formel InChI=1S/C9H13NO3/c10-8(11)6-4-1-2-5(3-4)7(6)9(12)13/h4-7H,1-3H2,(H2,10,11)(H,12,13)/t4-,5+,6+,7-/m1/s1
Formel vægt 183.09