Learn More
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
12638156
|
Epredia™
A78410023 |
Downdraft Extrctn Duct Adapter | |||||||
12648156
|
Epredia™
A78410024 |
Air System Duct Adapter Kit |
1370.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12683156
|
Epredia™
A78410026 |
Storage Bottle |
708.00 DKK
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12678156
|
Epredia™
A78410027 |
Waste Wax Drawer Kit | |||||||
12688156
|
Epredia™
A78410029 |
Monitor Clamp Kit | |||||||
12608346
|
Epredia™
B35210007 |
Basket Carrying Container |
208.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13470487
|
Epredia™
7411258 |
Vapor Filter (Charcoal) |
5270.00 DKK
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12764856
|
Epredia™
B9990612CS |
Potassium Permanganate Filter |
5205.00 DKK
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12658156
|
Epredia™
A78410025 |
Organized Basket, 222 cassette |
1470.00 DKK
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15594280
|
Epredia™
A82310038 |
Organized Basket, 300 cassette |
7255.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For use with Shandon Excelsior ES Tissue ProcessorSpot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Epredia™ Excelsior™ AS Tissue Processor Accessories