Learn More
Eutech™ 5.0 mS/cm KCl Calibration Solution
For conductivity calibration and standards, choose from our available Eutech™'s Conductivity Standards, Calibrations, and Solutions
Brand: Thermo Scientific Eutech 01X211272
Description
Eutech™ 5.0 mS/cm KCl Calibration Solution is used for calibrating conductivity probes for accurate measurement. Get quick and accurate conductivity measurements using Eutech™ conductivity meters and probes in the field, laboratories, and schools.
Features:
- Packed in 480ml durable plastic bottles
- Economical and convenient to use
Specifications
Calibration Solution | |
Conductivity Standard | |
480 mL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
015-07 | Certificate of Analysis | 27/12/2024 | 01X211272 |
015-03 | Certificate of Analysis | 27/12/2024 | 01X211272 |
015-09 | Certificate of Analysis | 27/12/2024 | 01X211272 |
015-08 | Certificate of Analysis | 27/12/2024 | 01X211272 |
015-02 | Certificate of Analysis | 27/12/2024 | 01X211272 |