Learn More
Potassium Chloride Solution (Conductivity Standard), 5.000μS/25°C, Chem Lab™
Brand: Chem Lab CL03.1139.0500
Quantity | 500 mL |
---|
This item is not returnable.
View return policy
Specifications
Odorless | |
Colorless | |
Soluble |
Liquid | |
±5 | |
500 mL |
Product Content Correction
Product Title
Potassium Chloride Solution (Conductivity Standard), 5.000μS/25°C, Chem Lab™ > Quantity: 500mL
Spot an opportunity for improvement?Share a Content Correction