Få mere at vide
Ethyl (Pentafluorobenzoyl)acetate, TRC
Artikelnummer. 30374972
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30374972 500 mg
30369892 50 mg
30370846 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30374972 Leverandør Toronto Research Chemicals Leverandørnr. TRCE818018500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3516-87-8
Molekylær formel C11H7O3F5
Molekylvægt (g/mol) 282.16
SMIL CCOC(=O)CC(=O)c1c(F)c(F)c(F)c(F)c1F
Tekniske data
Kemisk navn eller materiale Ethyl (Pentafluorobenzoyl)acetate
Mængde 500 mg
InChI formel InChI=1S/C11H7F5O3/c1-2-19-5(18)3-4(17)6-7(12)9(14)11(16)10(15)8(6)13/h2-3H2,1H3
Formel vægt 282.0315