missing translation for 'onlineSavingsMsg'
Få mere at vide

Ethyl 5-methylisoxazole-3-carboxylate, 95%, Thermo Scientific™

Artikelnummer. 11490743
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
25 g
Pakningsstørrelse:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
11490743 5 g 5g
11480743 1 g 1g
11400753 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11490743 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A13226.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 5-methylisoxazole-3-carboxylate, It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 3209-72-1
Molekylær formel C7H9NO3
Molekylvægt (g/mol) 155.15
MDL nummer MFCD00085080
InChI nøgle OCCIGHIQVMLYBZ-UHFFFAOYSA-N
Synonym ethyl 5-methylisoxazole-3-carboxylate, 3-isoxazolecarboxylic acid, 5-methyl-, ethyl ester, 5-methyl-isoxazole-3-carboxylic acid ethyl ester, 5-methylisoxazole-3-carboxylate ethyl, vitas-bb tbb000645, pubchem8773, 5-methylisoxazole-3-carboxylicacidethylester, acmc-1ctp0, ksc226e3b, 3-ethoxycarbonyl-5-methylisoxazole
PubChem CID 76677
IUPAC navn ethyl 5-methyl-1,2-oxazole-3-carboxylate
SMIL CCOC(=O)C1=NOC(C)=C1

Tekniske data

Kemisk navn eller materiale Ethyl 5-methylisoxazole-3-carboxylate
Smeltepunkt 27°C to 31°C
Kogepunkt 77°C to 79°C (0.5 mmHg)
Flammepunkt 109°C (228°F)
Mængde 5 g
Opløselighedsinformation It is insoluble in water.
Formel vægt 155.15
Procent renhed 95%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.