Få mere at vide
Alfa Aesar™ Ethyl 4,4-difluoroacetoacetate, 98%
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethyl 4,4-difluoroacetoacetate |
| CAS | 352-24-9 |
| Molekylær formel | C6H8F2O3 |
| MDL nummer | MFCD00039277 |
| Mængde | 1 g |
| FN nummer | UN1993 |
| Synonym | ethyl 4,4-difluoroacetoacetate, ethyl 4,4-difluoro-3-oxobutyrate, ethyl difluoroacetoacetate, butanoic acid, 4,4-difluoro-3-oxo-, ethyl ester, ethyl4,4-difluoro-3-oxobutyrate, ethyl 4,4-difluoro-3-oxo-butanoate, 4,4-difluoroacetoacetic acid ethyl ester, 4,4-difluoro-3-oxo-butyric acid ethyl ester, ethyl4,4-difluoro-3-oxobutanoate, pubchem21933 |
| InChI nøgle | CBDPWKVOPADMJC-UHFFFAOYSA-N |
| SMIL | CCOC(=O)CC(=O)C(F)F |
| IUPAC navn | ethyl 4,4-difluoro-3-oxobutanoate |
| Vis mere |