Learn More
Eppendorf™ epTIPS Box IVD
epT.I.P.S. pipette tips are designed to enable minimal insertion and ejection force and maximum tightness in system with Eppendorf pipettes.
488.70 DKK
Specifications
Filtered | Non-Filtered |
---|---|
Color | Colorless |
Autoclavable | Autoclavable |
Certifications/Compliance | 98/79 EC |
Clean Claims | Eppendorf Quality™ |
Product Code | Brand | Volume (Metric) | Color Code | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Color Code | Quantity | Price | Quantity & Availability | |||
17302930
|
Eppendorf™
0030076540 |
50 to 1000 μL | Blue | 480 Pk. |
488.70 DKK
Pack of 480 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||
Description
- epT.I.P.S.™ Box 2.0 (EU-IVD), Eppendorf Quality™
- Contamination-free transfer of trays to stable working box
- System optimized for use with multichannel pipettes
- Color-coded trays for simple identification of tips and pipette
- Tips can be attached to pipette from refill trays
- The trays and boxes can be autoclaved at 121 °C for subsequent use
- The pipette tips of the epT.I.P.S.™ product group with the respective product numbers are designed and constructed for low-contamination transfer of liquids, in particular for samples from the human body and for reagents within the scope of an in-vitro diagnostic application in order to allow the in-vitro diagnostic medical device to be used as intended. The above mentioned epT.I.P.S.™ are in-vitro diagnostic devices according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. They are intended exclusively for single use and for application by qualified staff.
Specifications
Non-Filtered | |
Autoclavable | |
Eppendorf Quality™ | |
Single-use | |
Polypropylene |
Colorless | |
98/79 EC | |
Consumables | |
epT.I.P.S. |
The pipette tips of the epT.I.P.S.™ product group with the respective product numbers are designed and constructed for low-contamination transfer of liquids, in particular for samples from the human body and for reagents within the scope of an in-vitro diagnostic application in order to allow the in-vitro diagnostic medical device to be used as intended. The above mentioned epT.I.P.S.™ are in-vitro diagnostic devices according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. They are intended exclusively for single use and for application by qualified staff.