missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Epstein Barr Virus Antibody (CS1, CS2, CS3, CS4), Novus Biologicals™
Mouse Monoclonal Antibody
1590.00 DKK - 3990.00 DKK
Tekniske data
| Antigen | Epstein Barr Virus |
|---|---|
| Klon | CS2, CS4, CS3, CS1 |
| Applikationer | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Konjugeret | Unconjugated |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
18298133
|
Novus Biologicals
NB120-17085-0.1ML |
0.1 mL |
1590.00 DKK
0.10ml |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
18179029
|
Novus Biologicals
NB120-17085 |
0.5 mL |
3990.00 DKK
0.50ml |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
Epstein Barr Virus Monoclonal specifically detects Epstein Barr Virus in Virus samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Tekniske data
| Epstein Barr Virus | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Immune System Diseases, Immunology, Virology Bacteria and Parasites | |
| Epstein Barr Virus, Human herpesvirus type 4, Hypothetical protein BALF1 | |
| IgG1 | |
| Tissue culture supernatant | |
| This antibody is specific to 60 kD latent membrane protein (LMP-1) encoded by the BNLF1 gene of EBV. Each clone reacts with different epitopes on the hydrophilic C-terminus of the cytoplasmic domain of LMP-1. This antibody stains strongly with EBV positive lymphoblastoid cell lines and EBV infected B cell immunoblasts in infectious mononucleosis. |
| CS2, CS4, CS3, CS1 | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Virus | |
| Recombinant fusion protein containing sequences of bacterial beta-galactosidase and EBV encoded latent membrane protein. | |
| Primary | |
| Store at 4°C. |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel