missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Epstein Barr Virus Antibody (CS1, CS2, CS3, CS4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-17085-0.1ML
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Epstein Barr Virus Monoclonal specifically detects Epstein Barr Virus in Virus samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Tekniske data
| Epstein Barr Virus | |
| Monoclonal | |
| Unconjugated | |
| Tissue culture supernatant, containing less than 0.1% sodium azide with Sodium Azide | |
| Mouse | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| Virus | |
| Supernatant |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CS2, CS4, CS3, CS1 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Epstein Barr Virus, Human herpesvirus type 4, Hypothetical protein BALF1 | |
| Recombinant fusion protein containing sequences of bacterial beta-galactosidase and EBV encoded latent membrane protein. | |
| 0.1 mL | |
| Immune System Diseases, Immunology, Virology Bacteria and Parasites | |
| This antibody is specific to 60 kD latent membrane protein (LMP-1) encoded by the BNLF1 gene of EBV. Each clone reacts with different epitopes on the hydrophilic C-terminus of the cytoplasmic domain of LMP-1. This antibody stains strongly with EBV positive lymphoblastoid cell lines and EBV infected B cell immunoblasts in infectious mononucleosis. | |
| Store at 4°C. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion