missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ ep Dualfilter T.I.P.S.™ SealMax (EU-IVD) Filter Tips
Eppendorf™ ep Dualfilter T.I.P.S.™ SealMax (EU-IVD) Filter Tips utilize a two-phase filter for protection of both sample and pipette
Brand: Eppendorf™ 0030078780
Additional Details : Weight : 1.11900kg
Description
- ep Dualfilter T.I.P.S. SealMax (EU-IVD) filter tips combine the unique and established two-phase filter for protection of both sample and pipette
- Utilizes an innovative liquid and moisture barrier
- Ideally suited for methods where preserving sample integrity and preventing cross-contamination are critical
- Reliable protection from liquids, aerosols, and biomolecules
- No PCR inhibiting additives
- Pipette is protected from aggressive liquids when over-aspiration occurs
- Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality
- Modern slim rack design with reclosable lid
- Improved sustainability with 20% less polypropylene consumed per rack
- Tips are designed and constructed for low-contamination transfer of liquids
- In particular for samples from the human body and for reagents within the scope of an in-vitro diagnostic application in order
- Allows the in-vitro diagnostic device to be used as intended
- ep Dualfilter T.I.P.S.™ SealMax are in-vitro diagnostic devices according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998
- They are intended exclusively for single use and for application by qualified staff
Specifications
2 to 200 μL | |
Sterile | |
Directive 98/79/EC | |
Rack | |
Consumables | |
Yellow | |
ep DualFilter T.I.P.S |
Filtered | |
55 mm | |
PCR Clean | |
960 tips | |
10 racks × 96 tips | |
Air-Cushion | |
Polypropylene |