missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Emtricitabine

CAS: 143491-57-0 | C8H10FN3O3S

737.00 DKK - 1800.00 DKK

Chemical Identifiers

CAS 143491-57-0
Molecular Formula C8H10FN3O3S
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15781789
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
462070010
1 g
737.00 DKK
1g
Please to purchase this item. Need a web account? Register with us today!
15791789
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
462070050
5 g
1800.00 DKK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

143491-57-0
C8H10FN3O3S

Specifications

143491-57-0
153622
C8H10FN3O3S
Emtricitabine
Solubility in water: soluble. Other solubilities: very soluble in methanol,very slightly soluble in acetonitrile
136°C to 140°C
247.24
Videos
SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Emtricitabine

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.