Learn More
Merck Immobilon™-P PVDF Membrane
Hydrophobic PVDF Transfer Membrane for western blotting
Brand:  Merck IPVH15150
1980.00 DKK valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10384511
  • 2165.00 DKK / Pack of 10
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
Description
Description
Immobilon™ PVDF membranes are offered in four types, each optimized for a different protein blotting application. Convenient blotting sandwiches feature pre-cut sheets of membrane and blotting filter paper. Immobilon™ PVDF membranes have high protein adsorption, so you won’t lose proteins during transfer or reprobing. The open pore structure makes it easy to access bound proteins and remove unbound probes. In addition, Immobilon™ PVDF membranes optimized for fluorescent blots dramatically increase signal-to-noise ratios for high sensitivity in quantitative, multiplexing applications.
Specifications
Specifications
15 cm
Optimized for binding proteins transferred from a variety of gel matricies
PVDF
15 cm
Sheet
0.45 μm