Learn More
Elma™ Lab Clean S10 Cleaning Solution
Safely cleans glass, metals including aluminum, ceramics and plastics. Elma™ Lab Clean S10 Cleaning Solution removes lime scale and lime soaps, oxides and tarnish, coolant emulsions, fingerprints, dust, mineral deposits and oils from instruments a
440.00 DKK - 1470.00 DKK
Specifications
Product Type | Ultrasonic Cleaning Solution |
---|---|
Form | Liquid |
pH | 2.5 |
Composition | Acidic |
Content And Storage | Keep between 43°F to 85°F, shelf life 2 years |
Alert:
Irritating to respiratory system and skin. Risk of serious damage to eyes.
Description
- Mildly acidic concentrate but safe for aluminum
- Dilute aqueous concentrate to 1% for use in an ultrasonic cleaner or to 0.5% in labware washers
- Foam-inhibited
- pH of concentrate: 2.5
- Biodegradable
- Available in 10 and 25L containers
Applications
- Cleans medical instruments, lab instruments and labware of glass, metal including aluminum, plastics and ceramics
Note
- Check magnesium alloys and acid-sensitive glassy materials for compatibility
Specifications
Ultrasonic Cleaning Solution | |
2.5 | |
Keep between 43°F to 85°F, shelf life 2 years |
Liquid | |
Acidic |
Safety and Handling
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P102-Keep out of reach of children.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.