missing translation for 'onlineSavingsMsg'
Få mere at vide

Eleclazine hydrochloride, MedChemExpress

Artikelnummer. 16489697
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16489697 5 mg
16493536 25 mg
16487168 10 mg
16451639 1 mg
16433197 50 mg
16417166 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16489697 Leverandør Medchem Express Leverandørnr. HY16738A

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Eleclazine (GS 6615) hydrochloride is a selective cardiac late sodium current inhibitor and a weak inhibitor of potassium current with IC50 value of <1 μM and approximately 14.2 μM, respectively.

Eleclazine hydrochloride shows concurrent protection against autonomically induced atrial premature beats, repolarization alternans and heterogeneity, and atrial fibrillation in porcine model. Eleclazine hydrochloride can be used to research cardiac arrhythmias.

Specifica

Kemisk navn eller materiale Eleclazine hydrochloride
CAS 1448754-43-5
Farve White
Renhedsgrad noter Research
Molekylær formel C21H17ClF3N3O3
Mængde 5 mg
Synonym GS 6615 hydrochloride
Opløselighedsinformation DMSO : ≥ 100 mg/mL (221.32 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMIL O=C1N(CC2=NC=CC=N2)CCOC3=CC=C(C4=CC=C(OC(F)(F)F)C=C4)C=C13.[H]Cl
Molekylvægt (g/mol) 451.83
Formel vægt 451.83
Procent renhed 99.27%
Grad Research
Fysisk form Solid
Vedi altri risultati Mostra meno risultati

Research purposes only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.