missing translation for 'onlineSavingsMsg'
Få mere at vide

Elastopar, TRC

Artikelnummer. 30378384
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378384 50 mg
30379581 100 mg
30373058 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30378384 Leverandør Toronto Research Chemicals Leverandørnr. TRCE32455050MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-80-9
Molekylær formel C7 H7 N3 O2
Molekylvægt (g/mol) 165.149
Synonym Aniline, N-methyl-N,p-dinitroso- (6CI,7CI,8CI), N-Methyl-N,4-dinitrosobenzenamine, Elastopar, Heat Pre, N,4-Dinitroso-N-methylaniline, N,4-Dinitroso-N-methylbenzenamine, N,p-Dinitroso-N-methylaniline, N-Methyl-N,4-dinitrosoaniline, Nitrosan K, Vulnoc DBM
IUPAC navn N-methyl-N-(4-nitrosophenyl)nitrous amide
SMIL CN(N=O)c1ccc(cc1)N=O

Specifications

Kemisk navn eller materiale N-Methyl-N,4-dinitrosobenzenamine
Mængde 50 mg
InChI formel InChI=1S/C7H7N3O2/c1-10(9-12)7-4-2-6(8-11)3-5-7/h2-5H,1H3
Formel vægt 165.0538
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.