Få mere at vide
Eidd 1931, TRC
Artikelnummer. 30134246
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134246 1 g
30135369 500 mg
30009368 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134246 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9858001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3258-02-4
Molekylær formel C9 H13 N3 O6
Molekylvægt (g/mol) 259.22
Synonym N4-Hydroxycytidine, N-Hydroxy-Cytidine, Nhc, (Z)-1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(Hydroxymethyl)Tetrahydrofuran-2-Yl)-4-(Hydroxyimino)-3,4-Dihydropyrimidin-2(1H)-One, Molnupiravir Impurity B, Eidd 1931
SMIL Oc[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=C\C(=N/O)\Nc2=O
Tekniske data
Kemisk navn eller materiale N-Hydroxycytidine
Mængde 1 g
InChI formel InChI=1S/C13H19N3O7/c1-6(2)12(19)22-5-7-9(17)10(18)11(23-7)16-4-3-8(15-21)14-13(16)20/h3-4,6-7,9-11,17-18,21H,5H2,1-2H3,(H,14,15,20)/t7-,9-,10-,11-/m1/s1
Formel vægt 259.0804