Få mere at vide
EHMT2-IN-2, MedChemExpress
Artikelnummer. 16467257
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
Pakningsstørrelse:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16467257 5 mg -
16440847 50 mg 50mg
16404548 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16467257 Leverandør Medchem Express Leverandørnr. HY111904
MedChemExpress EHMT2-IN-2 is a potent EHMT inhibitor, with IC50s of all <100 nM for EHMT1 peptide, EHMT2 peptide and cellular EHMT2. Used in the research of blood disease or cancer.
EHMT2-IN-2 is a potent EHMT inhibitor, with IC50s of all <100 nM for EHMT1 peptide, EHMT2 peptide and cellular EHMT2. Used in the research of blood disease or cancer.
Tekniske data
Kemisk navn eller materiale EHMT2-IN-2
CAS 2230850-47-0
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C21H22N6O
Mængde 5 mg
Opløselighedsinformation DMSO : 50 mg/mL (133.53 mM; Need ultrasonic)
SMIL CC1=CC(NC)=NC(NC2=CC=C(OC)C(C(N3)=C(C)C4=C3C=NC=C4)=C2)=N1
Molekylvægt (g/mol) 374.44
Formel vægt 374.44
Procent renhed 99.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation