Learn More
Invitrogen™ EGFR (ErbB1) [L858R] Recombinant Human Protein
The epidermal growth factor receptor is a receptor tyrosine kinases that signals in response to various growth factors.
Brand: Invitrogen™ PV4289
Additional Details : Weight : 0.00900kg
Description
Overexpression of EGFR has been linked to numerous types of cancer and EGFR is the target of both biological and small molecular therapeutics. The EGFR L858R mutation is implicated in epithelial tumor formation.
Specifications
NP_005219.2; L858R | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
1mg | |
RUO - research use only | |
EGFR (ErbB1) L858R | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
90.5kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
EGF Receptor (ErbB) Kinase Family | |
Recombinant | |
Baculovirus |