missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EGFR (ErbB1) [L858R] Recombinant Human Protein
The epidermal growth factor receptor is a receptor tyrosine kinases that signals in response to various growth factors.
13300.00 DKK - 57500.00 DKK
Specifications
Accession Number | NP_005219.2; L858R |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 90.5kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438593
|
Invitrogen™
PR7447A |
100μg |
13300.00 DKK
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10699624
|
Invitrogen™
PV4289 |
1mg |
57500.00 DKK
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Overexpression of EGFR has been linked to numerous types of cancer and EGFR is the target of both biological and small molecular therapeutics. The EGFR L858R mutation is implicated in epithelial tumor formation.
Specifications
NP_005219.2; L858R | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
EGF Receptor (ErbB) Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
90.5kDa | |
RUO - research use only | |
EGFR (ErbB1) L858R | |
Human | |
GST Tag | |
Frozen |