Learn More
Tenak™ ECO AluRack for Chest Freezers, 32 mm Box Height, with Boxes
ECO AluRack fits into any chest freezer and can be used with all standard cryoboxes.
700.00 DKK - 1130.00 DKK
Specifications
Includes | White cardboard boxes, 9 x 9 cell dividers |
---|---|
Description | Seawater resistant |
Material | Aluminum |
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 32 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12412590
|
Tenak™
TE11465B |
Boxes (15) |
955.00 DKK
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
12432580
|
Tenak™
TE11458B |
Boxes (8) |
700.00 DKK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
12432590
|
Tenak™
TE11466B |
Boxes (16) |
990.00 DKK
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
12452580
|
Tenak™
TE11460B |
Boxes (10) |
775.00 DKK
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
12452590
|
Tenak™
TE11468B |
Boxes (18) |
1060.00 DKK
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
12472580
|
Tenak™
TE11462B |
Boxes (12) |
840.00 DKK
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
12472590
|
Tenak™
TE11470B |
Boxes (20) |
1130.00 DKK
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
12492580
|
Tenak™
TE11464B |
Boxes (14) |
915.00 DKK
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
White cardboard boxes, 9 x 9 cell dividers | |
Aluminum | |
32 mm (Box) |
Seawater resistant | |
Chest Frseezers |