Learn More
Tenak™ ECO AluRack for Chest Freezers, 32 mm Box Height, with Boxes
ECO AluRack fits into any chest freezer and can be used with all standard cryoboxes.
Brand:  Tenak™ TE11468B
Additional Details : Weight : 1.60000kg
 View more versions of this product
Product Code. 12452590
  • 1060.00 DKK / Each
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
Description
Description
  • Made of seawater resistant aluminum
  • Fits into any freezer and can be used with all standard cryoboxes
  • Racks can be used with all freezer brands
  • Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
Specifications
White cardboard boxes, 9 x 9 cell dividers
Boxes (18)
Chest Frseezers
32 mm (Box)
Seawater resistant
Aluminum
140 x 140 x 648 mm