missing translation for 'onlineSavingsMsg'
Få mere at vide

EAI045, MedChemExpress

Artikelnummer. 16474256
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16474256 100 mg
16429506 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16474256 Leverandør Medchem Express Leverandørnr. HY100213

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress EAI045 is an allosteric and the fourth-generation inhibitor of mutant EGFR with IC50s of 1.9, 0.019, 0.19 and 0.002 μM for EGFR, EGFRL858R, EGFRT790M and EGFRL858R/T790M at 10 μM ATP, respectively.

EAI045 is an allosteric and the fourth-generation inhibitor of mutant EGFR with IC50s of 1.9, 0.019, 0.19 and 0.002 μM for EGFR, EGFRL858R, EGFRT790M and EGFRL858R/T790M at 10 μM ATP, respectively.

Specifications

Kemisk navn eller materiale EAI045
CAS 1942114-09-1
Farve off White
Renhedsgrad noter Research
Molekylær formel C19H14FN3O3S
Mængde 100 mg
Opløselighedsinformation DMSO : 100 mg/mL (260.82 mM; Need ultrasonic)
SMIL O=C(C(C1=C(O)C=CC(F)=C1)N2C(C(C=CC=C3)=C3C2)=O)NC4=NC=CS4
Molekylvægt (g/mol) 383.4
Formel vægt 383.4
Procent renhed 96.53%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.