missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
IBA Lifesciences pASG-IBA E. coli Expression Vector with No Secretion
Shop alle IBA Lifesciences produkter
Klik for at se tilgængelige muligheder
Mængde:
5 μg
Pakningsstørrelse:
5µg
Beskrivelse
- The pASG-IBA vectors are designed for expression of target proteins without affinity tag in E. coli.
- The vector carries an ampicillin resistance cassette for selection of transformed E. coli cells, F1 and ColE1 origin for a high plasmid copy number, and the inducible tetracycline promoter/operator for the regulated expression of proteins.
- The vectors can be combined with any E. coli strain since the tet-promoter works independently of the genetic background of E. coli.
- Expression of the recombinant protein is induced by addition of anhydrotetracycline, which is a derivative of tetracycline that does not exhibit antibiotic activity.
Tekniske data
Tekniske data
| Inducerende middel | Inducible with anhydrotetracycline |
| Markør Gene | Ampicillin |
| Promotor | Tet |
| Produkttype | Vector |
| Indhold og opbevaring | Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage |
| Protein tag | GST Tag |
| Mængde | 5 μg |
| Vektor | pASG-IBA25 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Spot an opportunity for improvement?Share a Content Correction