Få mere at vide
Dysprosium(III) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific™
910.46 DKK - 2313.47 DKK
Kemiske identifikatorer
| CAS | 14456-48-5 |
|---|---|
| Molekylær formel | Br3Dy |
| Molekylvægt (g/mol) | 402.212 |
| MDL nummer | MFCD00016069 |
| InChI nøgle | GBLDKMKYYYAAKD-UHFFFAOYSA-K |
| Synonym | dysprosium bromide, dysprosium tribromide, dysprosium iii bromide, dysprosium iii ; bromide, dysprosium 3+ tribromide, acmc-1bq18 |
| PubChem CID | 84453 |
| IUPAC navn | dysprosium(3+);tribromide |
| SMIL | [Br-].[Br-].[Br-].[Dy+3] |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
15467395
|
Thermo Scientific Alfa Aesar
047127.03 |
1g |
910.46 DKK
1g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
15477395
|
Thermo Scientific Alfa Aesar
047127.06 |
5g |
2313.47 DKK
5g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ampouled under argon
Kemiske identifikatorer
| 14456-48-5 | |
| 402.212 | |
| GBLDKMKYYYAAKD-UHFFFAOYSA-K | |
| 84453 | |
| [Br-].[Br-].[Br-].[Dy+3] |
| Br3Dy | |
| MFCD00016069 | |
| dysprosium bromide, dysprosium tribromide, dysprosium iii bromide, dysprosium iii ; bromide, dysprosium 3+ tribromide, acmc-1bq18 | |
| dysprosium(3+);tribromide |
Tekniske data
| Dysprosium(III) bromide | |
| 14456-48-5 | |
| Br3Dy | |
| dysprosium bromide, dysprosium tribromide, dysprosium iii bromide, dysprosium iii ; bromide, dysprosium 3+ tribromide, acmc-1bq18 | |
| [Br-].[Br-].[Br-].[Dy+3] | |
| 402.212 | |
| 402.23 | |
| Ampule |
| Ultra dry | |
| (metals basis) | |
| MFCD00016069 | |
| GBLDKMKYYYAAKD-UHFFFAOYSA-K | |
| dysprosium(3+);tribromide | |
| 84453 | |
| 99.99% | |
| Hygroscopic |
Sikkerhed og håndtering
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 238-443-9
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Anbefalet opbevaring : Ambient temperatures