Learn More
DWK Life Sciences DURAN™ Vacuum Receiver Adapter, bent, with 2 standard ground joints, and screw-on plastic hose connection
The receiver adapter is suitable for transferring the distillate from the condenser to the receiving flask.
Brand: DWK Life Sciences 241303401
455.05 DKK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.07800kg
Description
Part of a distillation assembly is, among other things, the receiver adapter, which is connected downstream at the end of the condenser and guides the distillate into the receiving flask.
- Tight closure due to standard ground joint
- Bent
- With two standard ground joint and screw-on plastic hose connection
- Fired-on, highly durable white print
Specifications
Autoclavable | |
8.6 mm | |
1 Ea. |
Distillation | |
Borosilicate Glass (3.3), PBT, Polypropylene, Silicone | |
NS 24/29 |