Learn More
DWK Life Sciences DURAN™ Vacuum Receiver Adapter, bent, with 2 standard ground joints, and screw-on plastic hose connection
The receiver adapter is suitable for transferring the distillate from the condenser to the receiving flask.
455.00 DKK - 540.00 DKK
Specifications
Description | Autoclavable |
---|---|
For Use With (Application) | Distillation |
Fits Tubing ID (Metric) | 8.6 mm |
Material | Borosilicate Glass (3.3), PBT, Polypropylene, Silicone |
Quantity | 1 Ea. |
Product Code | Brand | Thread Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | ||||||
10069940
|
DWK Life Sciences
241302105 |
NS 14/23 |
455.00 DKK
Each |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||||
10385191
|
DWK Life Sciences
241304603 |
NS 29/32 |
540.00 DKK
Each |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||||
10633792
|
DWK Life Sciences
241303401 |
NS 24/29 |
502.00 DKK
Each |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||||
Description
Part of a distillation assembly is, among other things, the receiver adapter, which is connected downstream at the end of the condenser and guides the distillate into the receiving flask.
- Tight closure due to standard ground joint
- Bent
- With two standard ground joint and screw-on plastic hose connection
- Fired-on, highly durable white print
Specifications
Autoclavable | |
8.6 mm | |
1 Ea. |
Distillation | |
Borosilicate Glass (3.3), PBT, Polypropylene, Silicone |