Learn More
DWK Life Sciences DURAN™ Separating Funnel stopcock Gilson, complete with SBW-J key
For attaching to DURAN™ separating funnel raw bodies.
Brand: DWK Life Sciences 286428407
3557.75 DKK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 2.50000kg
Description
DURAN™ Gilson separating funnel taps are components for the production of Gilson separating funnels with a particularly strong approach to reducing the risk of breakage.
- Separating funnel tap sleeve with SBW-J plug
- With a particularly strong approach to reducing the risk of breakage
- Glass articles for further processing
Specifications
2 mm | |
Borosilicate Glass (3.3), SBW-J, Polypropylene | |
12.5 NS | |
25/Pk. |
16 mm | |
No | |
1.8 mm | |
For attachment to DURAN™ separating funnel raw bodies; For the production of separating funnels according to Gilson |