Learn More
DWK Life Sciences DURAN™ Adapter, standard ground joint
The transition adapter connects two glass components with different standard ground joints.
Brand: DWK Life Sciences 241143203
201.40 DKK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.02600kg
Description
Transition adapters or reducers are glass components with two standard ground joints of different sizes. With the help of these adapters, glass components with different standard ground joints can be connected to each other.
- Tight closure due to standard ground joint
Specifications
1 Ea. | |
Connection of two glass components | |
Reduction Joint, Non-DIN Size | |
Socket NS 24/29, Cone NS 14/23 |