missing translation for 'onlineSavingsMsg'
Få mere at vide

DSP-4 hydrochloride, MedChemExpress

Artikelnummer. 16484856
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16484856 100 mg 100mg
16463808 10 mg -
16440187 50 mg -
16416537 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16484856 Leverandør Medchem Express Leverandørnr. HY103210

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress DSP-4 hydrochloride (Neurotoxin DSP 4 hydrochloride) is a highly selective neurotoxin and readily passes the blood-brain barrier with neurotoxic effects on noradrenergic neurons of adult and developing rats, can be used for the temporary selective degradation of the central and peripheral noradrenergic neurons, mainly those from the locus coeruleus (LC).

DSP-4 hydrochloride (Neurotoxin DSP 4 hydrochloride) is a highly selective neurotoxin and readily passes the blood-brain barrier with neurotoxic effects on noradrenergic neurons of adult and developing rats, can be used for the temporary selective degradation of the central and peripheral noradrenergic neurons, mainly those from the locus coeruleus (LC).

Tekniske data

Kemisk navn eller materiale DSP-4 hydrochloride
CAS 40616-75-9
Farve White
Renhedsgrad noter Research
Molekylær formel C11H16BrCl2N
Mængde 100 mg
Synonym Neurotoxin DSP 4 (hydrochloride)
Opløselighedsinformation DMSO : 125 mg/mL (399.28 mM; Need ultrasonic) ∣H2O : 50 mg/mL (159.71 mM; Need ultrasonic)
SMIL CCN(CCCl)CC1=CC=CC=C1Br.[H]Cl
Molekylvægt (g/mol) 313.06
Formel vægt 313.06
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.