missing translation for 'onlineSavingsMsg'
Få mere at vide

Droxinostat, MedChemExpress

Artikelnummer. 16446146
Encompass_Preferred
Change view
Click to view available options
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
16446146 100 mg
16435608 10 mg
16421777 50 mg
16408307 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16446146 Supplier Medchem Express Supplier No. HY13267

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Droxinostat(NS41080) is a selective inhibitor of HDAC3, HDAC6, and HDAC8 with IC50 of 16.9, 2.47 and 1.46 μM, respectively; > 8-fold selective against HDAC3 and no inhibition to HDAC1, 2, 4, 5, 7, 9, and 10.

Droxinostat(NS41080) is a selective inhibitor of HDAC3, HDAC6, and HDAC8 with IC50 of 16.9, 2.47 and 1.46 μM, respectively

Specifications

Kemisk navn eller materiale Droxinostat
CAS 99873-43-5
Farve White
Renhedsgrad noter Research
Molekylær formel C11H14ClNO3
Mængde 100 mg
Synonym NS 41080
Opløselighedsinformation DMSO : ≥ 150 mg/mL (615.54 mM)
SMIL ClC1=CC=C(C(C)=C1)OCCCC(NO)=O
Molekylvægt (g/mol) 243.69
Formel vægt 243.69
Procent renhed 98.78%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.