missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Donkey F(ab')2 anti-Mouse IgG (H+L) Secondary Antibody, Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NBP2-60648-0.5mg
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunomicroscopy
Tekniske data
| IgG (H+L) | |
| Polyclonal | |
| Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000 | |
| Donkey | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Flow Cytometry, Immunomicroscopy | |
| Unconjugated | |
| 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free | |
| This Donkey F(ab')2 anti-Mouse IgG (H+L) Secondary Antibody was developed against purified mouse IgG (H&L). | |
| 0.5 mg | |
| Adaptive Immunity, Immunology | |
| Mouse | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion