Få mere at vide
DO3A, TRC
Artikelnummer. 30137128
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137128 100 mg
30133667 50 mg
30009820 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137128 Leverandør Toronto Research Chemicals Leverandørnr. TRCD494885100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 217973-03-0
Molekylær formel C14H23N4O6 • 3(Na)
Molekylvægt (g/mol) 412.32
Synonym Trisodium 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetate, 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetic Acid Trisodium Salt
IUPAC navn sodium 2,2',2' '-(1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate
SMIL O=C(O[Na])CN1CCN(CC(O[Na])=O)CCN(CC(O[Na])=O)CCNCC1
Tekniske data
Kemisk navn eller materiale DO3A
Mængde 100 mg
InChI formel InChI=1S/C14H26N4O6.3Na/c19-12(20)9-16-3-1-15-2-4-17(10-13(21)22)6-8-18(7-5-16)11-14(23)24;;;/h15H,1-11H2,(H,19,20)(H,21,22)(H,23,24);;;/q;3*+1/p-3
Formel vægt 412.13