missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1043.29 DKK - 2973.37 DKK
Specifications
Antigen | DNA2 |
---|---|
Concentration | 0.29 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16898604
|
Proteintech
18727-1-AP-20UL |
20 μL |
1043.29 DKK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16888604
|
Proteintech
18727-1-AP-150UL |
150 μL |
2973.37 DKK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is specific to DNA2.
DNA2 is a conserved helicase/nuclease involved in themaintece of mitochondrial and nuclear DNA stability.Specifications
DNA2 | |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P51530, Q6ZQJ5 | |
1763, 327762 | |
Peptide | |
Primary | |
-20°C | |
DNA2 |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DNA2, DNA2L, KIAA0083 | |
Dna2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |