missing translation for 'onlineSavingsMsg'
Learn More

DNA2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18727-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16888604

  • 2973.37 DKK / 150µL
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is specific to DNA2.

DNA2 is a conserved helicase/nuclease involved in themaintece of mitochondrial and nuclear DNA stability.
Specifications

Specifications

DNA2
Polyclonal
Unconjugated
DNA2
DNA2, DNA2L, KIAA0083
Rabbit
Antigen Affinity Chromatography
RUO
1763, 327762
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P51530, Q6ZQJ5
Dna2
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers