Learn More
Thermo Scientific Chemicals DL-Methionine, 99% Product Code.: 11430333
Thermo Scientific Chemicals DL-Methionine, 99%
Product Code. p-7081465
250 g, 250g
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
Product Code. 11430333
Brand: Thermo Scientific Chemicals A11457.30
Request Bulk or Custom Format
CAS: 59-51-8 | C5H11NO2S | 149.21 g/mol
Production of volatile compounds related to the flavour of foods from the Strecker degradation of DL-methionine. Addition of methionine in appropriate amounts to these foods might be expected to improve protein value. Economically, DL-methionine would be preferable.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Production of volatile compounds related to the flavour of foods from the Strecker degradation of DL-methionine. Addition of methionine in appropriate amounts to these foods might be expected to improve protein value. Economically, DL-methionine would be preferable.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 59-51-8
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063096
InChI Key FFEARJCKVFRZRR-UHFFFAOYNA-N
Synonym dl-methionine, methionine, racemethionine, acimetion, banthionine, cynaron, lobamine, meonine, mertionin, metione
PubChem CID 876
ChEBI CHEBI:16811
SMILES CSCCC(N)C(O)=O
Specifications
Melting Point ∼272°C (decomposition)
Beilstein 636185
Merck Index 14,5975
Solubility Information Soluble in water.
Density 1.34
IUPAC Name 2-amino-4-methylsulfanylbutanoic acid
Quantity 250 g
Formula Weight 149.21
Percent Purity 99%
Chemical Name or Material DL-Methionine
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberM19K017 Certificate TypeCertificate of Analysis Date25/01/2024 Product CodeA11457.0B,A11457.0I,A11457.30
Lot Number10168212 Certificate TypeCertificate of Analysis Date10/01/2023 Product CodeA11457.0B,A11457.0I,A11457.30
Lot Number10243902 Certificate TypeCertificate of Analysis Date13/10/2022 Product CodeA11457.0B,A11457.0I,A11457.30
Lot NumberP07I050 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeA11457.0B,A11457.0I,A11457.30
Lot Number10158665 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA11457.0B,A11457.0I,A11457.30
RUO – Research Use Only
Product Content Correction
Product Title
Thermo Scientific Chemicals DL-Methionine, 99% >