Få mere at vide
Diquat Monopyridone Bromide, TRC
Artikelnummer. 30378879
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378879 25 mg
30371330 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30378879 Leverandør Toronto Research Chemicals Leverandørnr. TRCD49291025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 54016-01-2
Molekylær formel C12H11BrN2O
Molekylvægt (g/mol) 279.13
Synonym 6,7-Dihydro-4-oxo-dipyrido[1,2-a:2',1'-c]pyrazin-8-ium Bromide
SMIL O=C1C=CC=C2C3=[N+](C=CC=C3)CCN21.[Br-]
Tekniske data
Kemisk navn eller materiale Diquat Monopyridone Bromide
Mængde 25 mg
InChI formel InChI=1S/C12H11N2O.BrH/c15-12-6-3-5-11-10-4-1-2-7-13(10)8-9-14(11)12;/h1-7H,8-9H2;1H/q+1;/p-1
Formel vægt 278.0055