Learn More
Fistreem International™ Dip Tube (Boiler), Loughborough Water Still
Specifically designed for a Loughborough water still. Fistreem International™ Dip Tube (Boiler), Loughborough Water Still directs incoming cold replacement water to the bottom of the boiler.
Brand: Fistreem International™ ALB/2/8
Additional Details : Weight : 5.00000kg
Description
- For Loughborough water still.
- Directs incoming cold replacement water to the bottom of the boiler.
- Includes silicone rubber tip.
Specifications
Silicone Rubber Tip | |
WS/8 Water Still | |
Dip Tube | |
Silicone Rubber |