Learn More
Fistreem International™ Dip Tube (Boiler), Loughborough Water Still
Specifically designed for a Loughborough water still. Fistreem International™ Dip Tube (Boiler), Loughborough Water Still directs incoming cold replacement water to the bottom of the boiler.
472.00 DKK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
13156000
|
Fistreem International™
ALB/2/4 |
WS/4 Water Still |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13166000
|
Fistreem International™
ALB/2/8 |
WS/8 Water Still |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- For Loughborough water still.
- Directs incoming cold replacement water to the bottom of the boiler.
- Includes silicone rubber tip.
Specifications
Dip Tube | |
Silicone Rubber Tip |