Få mere at vide
Dimetofrine, TRC
Artikelnummer. 30369594
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30369594 10 mg
30368949 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30369594 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47971510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22950-29-4
Molekylær formel C11 H17 N O4
Molekylvægt (g/mol) 227.26
Synonym 4-Hydroxy-3,5-dimethoxy-α-[(methylamino)methyl]benzenemethanol
IUPAC navn 4-[1-hydroxy-2-(methylamino)ethyl]-2,6-dimethoxyphenol
SMIL CNCC(O)c1cc(OC)c(O)c(OC)c1
Tekniske data
Kemisk navn eller materiale Dimetofrine
Mængde 10 mg
InChI formel InChI=1S/C11H17NO4/c1-12-6-8(13)7-4-9(15-2)11(14)10(5-7)16-3/h4-5,8,12-14H,6H2,1-3H3
Formel vægt 227.1158