Få mere at vide
Dihydrodiol Ibrutinib, TRC
Artikelnummer. 30179294
Skift visning
Mængde:
2.5 mg
5 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30179294 2.5 mg 2.5mg
30179438 5 mg -
30178994 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179294 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4502502.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1654820-87-7
Molekylær formel C25H26N6O4
Molekylvægt (g/mol) 474.51
Synonym 1-[3-[4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]piperidin-1-yl]-2,3-dihydroxypropan-1-one, 1-[(3R)-3-[4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-1-piperidinyl]-2,3-dihydroxy-1-propanone, PCI 45227
SMIL NC1=C2C(N([C@@H]3CCCN(C(C(O)CO)=O)C3)N=C2C4=CC=C(OC5=CC=CC=C5)C=C4)=NC=N1
Tekniske data
Kemisk navn eller materiale Dihydrodiol Ibrutinib
Mængde 2.5 mg
InChI formel InChI=1S/C25H26N6O4/c26-23-21-22(16-8-10-19(11-9-16)35-18-6-2-1-3-7-18)29-31(24(21)28-15-27-23)17-5-4-12-30(13-17)25(34)20(33)14-32/h1-3,6-11,15,17,20,32-33H,4-5,12-14H2,(H2,26,27,28)/t17-,20?/m1/s1