Få mere at vide
Dihydroartemisinin Diketo Aldehyde, TRC
Artikelnummer. 30011546
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011546 25 mg
30005917 100 mg
30004621 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011546 Leverandør Toronto Research Chemicals Leverandørnr. TRCO84848025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1093625-96-7
Molekylær formel C14 H22 O3
Molekylvægt (g/mol) 238.32
Synonym (2S,3R,6RS)-2-(3-Oxobutyl)-3-methyl-6-[(R)-2-propanal]cyclohexanoneMixture of Diastereomers, (αR,3S,4R)-α,4-Dimethyl-2-oxo-3-(3-oxobutyl)cyclohexaneacetaldehyde
IUPAC navn (2R)-2-[(3S,4R)-4-methyl-2-oxo-3-(3-oxobutyl)cyclohexyl]propanal
SMIL C[C@@H](C=O)C1CC[C@@H](C)[C@H](CCC(=O)C)C1=O
Tekniske data
Kemisk navn eller materiale (2S,3R,6RS)-2-(3-Oxobutyl)-3-methyl-6-[(R)-2-propanal]cyclohexanone (mixture of diastereomers)
Mængde 25 mg
InChI formel InChI=1S/C14H22O3/c1-9-4-6-13(10(2)8-15)14(17)12(9)7-5-11(3)16/h8-10,12-13H,4-7H2,1-3H3/t9-,10+,12+,13?/m1/s1
Formel vægt 238.1569