Få mere at vide
Diflorasone 17-propionate-21-mesylate, TRC
Artikelnummer. 30137848
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137848 100 mg
30132701 50 mg
30008808 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137848 Leverandør Toronto Research Chemicals Leverandørnr. TRCD445513100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 84509-92-2
Molekylær formel C26H34F2O8S
Molekylvægt (g/mol) 544.61
Synonym (6α,11β,16β)-6,9-Difluoro-11-hydroxy-16-methyl-21-[(methylsulfonyl)oxy]-17-(1-oxopropoxy)pregna-1,4-diene-3,20-dione
IUPAC navn (6S,8S,9R,10S,11S,13S,14S,16S,17R)-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-17-(2-((methylsulfonyl)oxy)acetyl)-3-oxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthren-17-yl propionate
SMIL O=C([C@]([C@]([C@@]1([H])C2)(C[C@H](O)[C@](F)([C@]3(C=C4)C)[C@@]1([H])C[C@H](F)C3=CC4=O)C)([C@H]2C)OC(CC)=O)CO[S](=O)(C)=O
Tekniske data
Kemisk navn eller materiale Diflorasone 17-Propionate-21-mesylate
Mængde 100 mg
InChI formel InChI=1S/C26H34F2O8S/c1-6-22(32)36-26(21(31)13-35-37(5,33)34)14(2)9-16-17-11-19(27)18-10-15(29)7-8-23(18,3)25(17,28)20(30)12-24(16,26)4/h7-8,10,14,16-17,19-20,30H,6,9,11-13H2,1-5H3/t14-,16-,17-,19-,20-,23-,24-,25-,26-/m0/s1
Formel vægt 544.19