missing translation for 'onlineSavingsMsg'
Learn More
Diethylene Glycol Monoethyl Ether, ReagentPlus™ 99%, Honeywell™ Product Code.: 15668420

Diethylene Glycol Monoethyl Ether, ReagentPlus™ 99%, Honeywell™

Product Code. 15668420 Shop All Honeywell Research Chemicals Products
4 L, Glass bottle
Click to view available options
Quantity:
100 mL
1 L
4 L
20 L
Packaging:
Glass bottle
Stainless steel drum
This item is not returnable. View return policy

Product Code. 15668420

Brand: Honeywell Research Chemicals 5376164L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-90-0
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.175
MDL Number MFCD00002872
InChI Key XXJWXESWEXIICW-UHFFFAOYSA-N
Synonym diethylene glycol monoethyl ether, 2-2-ethoxyethoxy ethanol, carbitol, transcutol, ethoxy diglycol, dioxitol, ethyl carbitol, ethyl digol, carbitol solvent, ethanol, 2-2-ethoxyethoxyShow MoreShow Less
PubChem CID 8146
ChEBI CHEBI:40572
IUPAC Name 2-(2-ethoxyethoxy)ethanol
SMILES CCOCCOCCO

Specifications

Density 0.999g/mL at 25°C
Packaging Glass bottle
Linear Formula C2H5OCH2CH2OCH2CH2OH
Refractive Index 1.427
Quantity 4 L
UN Number NONH for all modes of transport
Beilstein 1736441
Vapor Density 4.63 (vs air)
Formula Weight 134.17g/mol
Percent Purity 99%
Chemical Name or Material Diethylene glycol monoethyl ether
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethylene Glycol Monoethyl Ether, ReagentPlus™ 99%, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.